Merck
  • Glutamine Metabolism Controls Stem Cell Fate Reversibility and Long-Term Maintenance in the Hair Follicle.
Glutamine Metabolism Controls Stem Cell Fate Reversibility and Long-Term Maintenance in the Hair Follicle.
Cell metabolism (2020-09-10)
Christine S Kim, Xiaolei Ding, Kira Allmeroth, Leah C Biggs, Olivia I Kolenc, Nina L'Hoest, Carlos Andrés Chacón-Martínez, Christian Edlich-Muth, Patrick Giavalisco, Kyle P Quinn, Martin S Denzel, Sabine A Eming, Sara A Wickström
ABSTRACT
Stem cells reside in specialized niches that are critical for their function. Upon activation, hair follicle stem cells (HFSCs) exit their niche to generate the outer root sheath (ORS), but a subset of ORS progeny returns to the niche to resume an SC state. Mechanisms of this fate reversibility are unclear. We show that the ability of ORS cells to return to the SC state requires suppression of a metabolic switch from glycolysis to oxidative phosphorylation and glutamine metabolism that occurs during early HFSC lineage progression. HFSC fate reversibility and glutamine metabolism are regulated by the mammalian target of rapamycin complex 2 (mTORC2)-Akt signaling axis within the niche. Deletion of mTORC2 results in a failure to re-establish the HFSC niche, defective hair follicle regeneration, and compromised long-term maintenance of HFSCs. These findings highlight the importance of spatiotemporal control of SC metabolic states in organ homeostasis.
Sigma-Aldrich
VEGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Methoxyamine hydrochloride, 98%
Sigma-Aldrich
O-Phosphorylethanolamine
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
SIGMAFAST 3,3′-Diaminobenzidine tablets, tablet, To prepare 1 mL
Swagelok® Front Ferrule, Swagelok® 203-1, stainless steel, 1/8 in. Swagelok
Sigma-Aldrich
Sodium dichloroacetate, 98%
Sigma-Aldrich
Citric acid-2,2,4,4-d4, 98 atom % D, 98% (CP)
Sigma-Aldrich
BPTES, ≥95% (HPLC)