Skip to Content
Merck
All Photos(5)

Key Documents

226904

Sigma-Aldrich

Methoxyamine hydrochloride

98%

Synonym(s):

O-Methylhydroxylamine hydrochloride, Methoxylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3ONH2 · HCl
CAS Number:
Molecular Weight:
83.52
Beilstein:
3589723
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

151-154 °C (lit.)

solubility

alcohol: soluble(lit.)
water: soluble(lit.)

SMILES string

Cl.CON

InChI

1S/CH5NO.ClH/c1-3-2;/h2H2,1H3;1H

InChI key

XNXVOSBNFZWHBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methoxyamine hydrochloride combines with human antithrombin III-thrombin complex to form an inactive thrombin.

Application

Methoxyamine hydrochloride was used as a reagent in the preparation of O-methyl oximes. It was also used in the synthesis of O-methyl oximes from aldehydes or ketones.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edouard Godineau et al.
Organic letters, 8(21), 4871-4874 (2006-10-06)
[reaction: see text] A formal [2+2+2] process has been devised that allows the stereocontrolled formation of ring-fused piperidines from allylsilanes possessing an oxime moiety. The cascade involves an intermolecular radical addition of an alpha-iodoacetate onto an allylsilane double bond, which
Tetrahedron Letters, 47, 6045-6045 (2006)
M O Longas et al.
The Biochemical journal, 189(3), 481-489 (1980-09-01)
1. Cleavage of the human antithrombin III--thrombin complex with [14C]methoxyamine hydrochloride results in inactive thrombin and 14C-labelled antithrombin III. 2. Discontinuous polyacrylamide-gel electrophoresis of the reduced dissociation fragments of the complex in the presence of sodium dodecyl sulphate reveals two
Jean-Philippe Mevy et al.
Plants (Basel, Switzerland), 9(9) (2020-09-10)
Global change scenarios in the Mediterranean basin predict a precipitation reduction within the coming hundred years. Therefore, increased drought will affect forests both in terms of adaptive ecology and ecosystemic services. However, how vegetation might adapt to drought is poorly
Chunmei Geng et al.
Scientific reports, 10(1), 700-700 (2020-01-22)
Depression is the most common disabling psychiatric disease, with a high prevalence and mortality. Chronic unpredictable mild stress (CUMS) is a well-accepted method used to mimic clinical depression. Recent evidence has consistently suggested that the cumulative effects of CUMS could

Articles

DNA damage and repair mechanism is vital for maintaining DNA integrity. Damage to cellular DNA is involved in mutagenesis, the development of cancer among others.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service