Merck
A potent, versatile disulfide-reducing agent from aspartic acid.
Journal of the American Chemical Society (2012-02-23)
John C Lukesh, Michael J Palte, Ronald T Raines
ABSTRACT
Dithiothreitol (DTT) is the standard reagent for reducing disulfide bonds between and within biological molecules. At neutral pH, however, >99% of DTT thiol groups are protonated and thus unreactive. Herein, we report on (2S)-2-amino-1,4-dimercaptobutane (dithiobutylamine or DTBA), a dithiol that can be synthesized from l-aspartic acid in a few high-yielding steps that are amenable to a large-scale process. DTBA has thiol pK(a) values that are ~1 unit lower than those of DTT and forms a disulfide with a similar E°' value. DTBA reduces disulfide bonds in both small molecules and proteins faster than does DTT. The amino group of DTBA enables its isolation by cation-exchange and facilitates its conjugation. These attributes indicate that DTBA is a superior reagent for reducing disulfide bonds in aqueous solution.
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, ~1 M in H2O