Merck
  • Sequential role of RAD51 paralog complexes in replication fork remodeling and restart.
Sequential role of RAD51 paralog complexes in replication fork remodeling and restart.
Nature communications (2020-07-17)
Matteo Berti, Federico Teloni, Sofija Mijic, Sebastian Ursich, Jevgenij Fuchs, Maria Dilia Palumbieri, Jana Krietsch, Jonas A Schmid, Edwige B Garcin, Stéphanie Gon, Mauro Modesti, Matthias Altmeyer, Massimo Lopes
ABSTRACT
Homologous recombination (HR) factors were recently implicated in DNA replication fork remodeling and protection. While maintaining genome stability, HR-mediated fork remodeling promotes cancer chemoresistance, by as-yet elusive mechanisms. Five HR cofactors - the RAD51 paralogs RAD51B, RAD51C, RAD51D, XRCC2 and XRCC3 - recently emerged as crucial tumor suppressors. Albeit extensively characterized in DNA repair, their role in replication has not been addressed systematically. Here, we identify all RAD51 paralogs while screening for modulators of RAD51 recombinase upon replication stress. Single-molecule analysis of fork progression and architecture in isogenic cellular systems shows that the BCDX2 subcomplex restrains fork progression upon stress, promoting fork reversal. Accordingly, BCDX2 primes unscheduled degradation of reversed forks in BRCA2-defective cells, boosting genomic instability. Conversely, the CX3 subcomplex is dispensable for fork reversal, but mediates efficient restart of reversed forks. We propose that RAD51 paralogs sequentially orchestrate clinically relevant transactions at replication forks, cooperatively promoting fork remodeling and restart.
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder
Sigma-Aldrich
Ribonuclease A from bovine pancreas, Type I-AS, 50-100 Kunitz units/mg protein
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Mirin, ≥98% (HPLC), powder
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Millipore
Microcon® DNA Fast Flow Centrifugal Filters, sample volume 0.5 mL, Ultracel® regenerated cellulose membrane (low binding)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Hydroxyurea, 98%, powder