Merck
  • Site-Dependent Cysteine Lipidation Potentiates the Activation of Proapoptotic BAX.
Site-Dependent Cysteine Lipidation Potentiates the Activation of Proapoptotic BAX.
Cell reports (2020-03-12)
Daniel T Cohen, Thomas E Wales, Matthew W McHenry, John R Engen, Loren D Walensky
ABSTRACT
BCL-2 family proteins converge at the mitochondrial outer membrane to regulate apoptosis and maintain the critical balance between cellular life and death. This physiologic process is essential to organism homeostasis and relies on protein-protein and protein-lipid interactions among BCL-2 family proteins in the mitochondrial lipid environment. Here, we find that trans-2-hexadecenal (t-2-hex), previously implicated in regulating BAX-mediated apoptosis, does so by direct covalent reaction with C126, which is located on the surface of BAX at the junction of its α5/α6 core hydrophobic hairpin. The application of nuclear magnetic resonance spectroscopy, hydrogen-deuterium exchange mass spectrometry, specialized t-2-hex-containing liposomes, and BAX mutational studies in mitochondria and cells reveals structure-function insights into the mechanism by which covalent lipidation at the mitochondria sensitizes direct BAX activation. The functional role of BAX lipidation as a control point of mitochondrial apoptosis could provide a therapeutic strategy for BAX modulation by chemical modification of C126.
Sigma-Aldrich
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97%
Sigma-Aldrich
Propargyl bromide solution, 80 wt. % in toluene, contains 0.3% magnesium oxide as stabilizer
Sigma-Aldrich
trans-2-Decenal, ≥95.0% (GC)
Sigma-Aldrich
trans-2-Pentenal, 95%
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
8-Aminonaphthalene-1,3,6-trisulfonic acid disodium salt, BioReagent, suitable for fluorescence, ≥90% (CE)
Sigma-Aldrich
trans-2-Octenal, technical grade, 94%
Sigma-Aldrich
Di-tert-butyl hydrazodiformate, 97%
Sigma-Aldrich
trans-2-Undecenal, ≥95%, stabilized, FG
Sigma-Aldrich
trans-2-Hexen-1-al, 98%
Sigma-Aldrich
trans-2-Nonenal, 97%
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
Protease from Aspergillus saitoi, Type XIII, ≥0.6 unit/mg solid
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
trans-2-Dodecenal, ≥93%, stabilized, FG