Skip to Content
Merck
All Photos(2)

Documents

269956

Sigma-Aldrich

trans-2-Octenal

technical grade, 94%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CHCHO
CAS Number:
Molecular Weight:
126.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

vapor density

>1 (vs air)

Assay

94%

form

liquid

refractive index

n20/D 1.45 (lit.)

bp

84-86 °C/19 mmHg (lit.)

density

0.846 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)C(\[H])=C(/[H])CCCCC

InChI

1S/C8H14O/c1-2-3-4-5-6-7-8-9/h6-8H,2-5H2,1H3/b7-6+

InChI key

LVBXEMGDVWVTGY-VOTSOKGWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The rate constant for the gas-phase reaction of the NO(3) radical with trans-2-octenal was studied using absolute rate method. The fungal volatile organic compound trans-2-octenal, caused locomotory defects and changes in green fluorescent protein (GFP) and antigen-labeled dopaminergic neurons in adult Drosophila melanogaster.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michaela Bodner et al.
Journal of chemical ecology, 38(5), 547-556 (2012-05-15)
The secretions from serial defensive glands of the Austrian diplopod Allajulus dicentrus (Julidae, Cylindroiulini) were extracted and analyzed by means of gas chromatography - mass spectrometry. In adults, 13 components from two chemical classes were detected: 1) The common juliform
K Hasegawa et al.
Biochimica et biophysica acta, 962(3), 371-376 (1988-10-14)
The structures of fluorescent products formed in the reaction of methyl linoleate hydroperoxides with adenine, FeSO4 and ascorbic acid were investigated to elucidate the mechanism of interaction. The fluorescent products consisted of at least four major components (I-IV), which could
M Alaiz et al.
Biological & pharmaceutical bulletin, 17(1), 51-57 (1994-01-01)
It is known that macrophages recognize and take up oxidized low density lipoprotein (LDL) and this macrophage recognition has been suggested to be due to modification of the lysine (Lys) residues of apoprotein B (apo B). In order to determine
Alessandro Genovese et al.
Molecules (Basel, Switzerland), 25(8) (2020-04-29)
In this study, the influence of phenolic compounds on the sensory scores attributed to extra virgin olive oil (EVOO) by panel test was investigated. Two model olive oils (MOOs) with identical concentrations of volatile compounds, differing only in the amount
Vincent Harraca et al.
BMC biology, 8, 121-121 (2010-09-11)
Abdominal wounding by traumatic insemination and the lack of a long distance attraction pheromone set the scene for unusual sexual signalling systems. Male bed bugs (Cimex lectularius) mount any large, newly fed individual in an attempt to mate. Last instar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service