Merck
  • Cholesterol Stabilizes TAZ in Hepatocytes to Promote Experimental Non-alcoholic Steatohepatitis.
Cholesterol Stabilizes TAZ in Hepatocytes to Promote Experimental Non-alcoholic Steatohepatitis.
Cell metabolism (2020-04-08)
Xiaobo Wang, Bishuang Cai, Xiaoming Yang, Oluwatoni O Sonubi, Ze Zheng, Rajasekhar Ramakrishnan, Hongxue Shi, Luca Valenti, Utpal B Pajvani, Jaspreet Sandhu, Rodney E Infante, Arun Radhakrishnan, Douglas F Covey, Kun-Liang Guan, Jochen Buck, Lonny R Levin, Peter Tontonoz, Robert F Schwabe, Ira Tabas
ABSTRACT
Incomplete understanding of how hepatosteatosis transitions to fibrotic non-alcoholic steatohepatitis (NASH) has limited therapeutic options. Two molecules that are elevated in hepatocytes in human NASH liver are cholesterol, whose mechanistic link to NASH remains incompletely understood, and TAZ, a transcriptional regulator that promotes fibrosis but whose mechanism of increase in NASH is unknown. We now show that increased hepatocyte cholesterol upregulates TAZ and promotes fibrotic NASH. ASTER-B/C-mediated internalization of plasma membrane cholesterol activates soluble adenylyl cyclase (sAC; ADCY10), triggering a calcium-RhoA-mediated pathway that suppresses β-TrCP/proteasome-mediated TAZ degradation. In mice fed with a cholesterol-rich NASH-inducing diet, hepatocyte-specific silencing of ASTER-B/C, sAC, or RhoA decreased TAZ and ameliorated fibrotic NASH. The cholesterol-TAZ pathway is present in primary human hepatocytes, and associations among liver cholesterol, TAZ, and RhoA in human NASH liver are consistent with the pathway. Thus, hepatocyte cholesterol contributes to fibrotic NASH by increasing TAZ, suggesting new targets for therapeutic intervention.
Sigma-Aldrich
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
D-(−)-Fructose, ≥99% (HPLC), BioXtra
Avanti
14:0 PC (DMPC), 1,2-dimyristoyl-sn-glycero-3-phosphocholine, powder
Avanti
14:0 PC (DMPC), 1,2-dimyristoyl-sn-glycero-3-phosphocholine, chloroform
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests
CellCrown inserts, 6 well plate inserts with 1.0 μm polycarbonate filter, sterile