Merck
  • Environmental arginine controls multinuclear giant cell metabolism and formation.
Environmental arginine controls multinuclear giant cell metabolism and formation.
Nature communications (2020-01-24)
Julia S Brunner, Loan Vulliard, Melanie Hofmann, Markus Kieler, Alexander Lercher, Andrea Vogel, Marion Russier, Johanna B Brüggenthies, Martina Kerndl, Victoria Saferding, Birgit Niederreiter, Alexandra Junza, Annika Frauenstein, Carina Scholtysek, Yohei Mikami, Kristaps Klavins, Gerhard Krönke, Andreas Bergthaler, John J O'Shea, Thomas Weichhart, Felix Meissner, Josef S Smolen, Paul Cheng, Oscar Yanes, Jörg Menche, Peter J Murray, Omar Sharif, Stephan Blüml, Gernot Schabbauer
ABSTRACT
Multinucleated giant cells (MGCs) are implicated in many diseases including schistosomiasis, sarcoidosis and arthritis. MGC generation is energy intensive to enforce membrane fusion and cytoplasmic expansion. Using receptor activator of nuclear factor kappa-Β ligand (RANKL) induced osteoclastogenesis to model MGC formation, here we report RANKL cellular programming requires extracellular arginine. Systemic arginine restriction improves outcome in multiple murine arthritis models and its removal induces preosteoclast metabolic quiescence, associated with impaired tricarboxylic acid (TCA) cycle function and metabolite induction. Effects of arginine deprivation on osteoclastogenesis are independent of mTORC1 activity or global transcriptional and translational inhibition. Arginine scarcity also dampens generation of IL-4 induced MGCs. Strikingly, in extracellular arginine absence, both cell types display flexibility as their formation can be restored with select arginine precursors. These data establish how environmental amino acids control the metabolic fate of polykaryons and suggest metabolic ways to manipulate MGC-associated pathologies and bone remodelling.
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Freund′s Adjuvant, Complete, cell suspension
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Histopaque®-1077, sterile-filtered, density: 1.077
Sigma-Aldrich
Leukocyte Acid Phosphatase (TRAP) Kit, Kit formulated with all liquid reagents
Sigma-Aldrich
Ammonium chloride-ammonium hydroxide buffer solution, in H2O
Sigma-Aldrich
Anti-Arginase-1 Antibody, from chicken
Sigma-Aldrich
Aquatex®, (aqueous mounting agent) for microscopy
Sigma-Aldrich
Accutase® solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Histopaque®-1119, sterile-filtered, density: 1.119