Merck
  • Microtubule-Actomyosin Mechanical Cooperation during Contact Guidance Sensing.
Microtubule-Actomyosin Mechanical Cooperation during Contact Guidance Sensing.
Cell reports (2018-10-12)
Erdem D Tabdanov, Vikram Puram, Alexander Zhovmer, Paolo P Provenzano
ABSTRACT
Cancer cell migration through and away from tumors is driven in part by migration along aligned extracellular matrix, a process known as contact guidance (CG). To concurrently study the influence of architectural and mechanical regulators of CG sensing, we developed a set of CG platforms. Using flat and nanotextured substrates with variable architectures and stiffness, we show that CG sensing is regulated by substrate stiffness and define a mechanical role for microtubules and actomyosin-microtubule interactions during CG sensing. Furthermore, we show that Arp2/3-dependent lamellipodia dynamics can compete with aligned protrusions to diminish the CG response and define Arp2/3- and Formins-dependent actin architectures that regulate microtubule-dependent protrusions, which promote the CG response. Thus, our work represents a comprehensive examination of the physical mechanisms influencing CG sensing.
Sigma-Aldrich
4-Nitro-3-(trifluoromethyl)phenol, 99% (GC)
Sigma-Aldrich
(+)-Biotin N-hydroxysuccinimide ester, ≥98% (HPLC), powder
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
(-)-Blebbistatin, The active enantiomer of (±)-Blebbistatin that accounts for the inhibitory activity towards ATPase and myosin II-dependent cellular processes.
Sigma-Aldrich
SMIFH2, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane
Supelco
Paclitaxel, Pharmaceutical Secondary Standard; Certified Reference Material