Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
glass microfiber filtration funnel
Products
Product Category
funnels (13)
holders (12)
Manufacturer
Capacity (mL)
Diameter (mm)
Volume (ml)
Plate Material
Package Size
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
WHA1950009
Whatman
®
glass microfiber filtration funnel,
Filter Funnel, 3-piece, volume 200 mL, diam. 90 mm
Expand
View Pricing
WHA1950004
Whatman
®
glass microfiber filtration funnel,
Filter Funnel, 3-piece, volume 36 mL, diam. 47 mm
Expand
View Pricing
WHA1950002
Whatman
®
glass microfiber filtration funnel,
Filter Funnel, 3-piece, volume 16 mL, diam. 25 mm
Expand
View Pricing
WHA1950007
Whatman
®
glass microfiber filtration funnel,
Filter Funnel, 3-piece, volume 115 mL, diam. 70 mm
Expand
View Pricing
WHA1950114
Whatman
®
glass microfiber filtration funnel,
PTFE plate, diam. 47 mm
Expand
View Pricing
WHA1950109
Whatman
®
glass microfiber filtration funnel,
polypropylene plate, diam. 90 mm
Expand
View Pricing
WHA1950012
Whatman
®
glass microfiber filtration funnel,
Filter Funnel, 3-piece, volume 530 mL, diam. 125 mm
Expand
View Pricing
WHA1950209
Whatman
®
glass microfiber filtration funnel,
capacity 200 mL , reservoir, diam. 90 mm
Expand
View Pricing
WHA1950117
Whatman
®
glass microfiber filtration funnel,
PTFE plate, diam. 70 mm
Expand
View Pricing
WHA1950017
Whatman
®
glass microfiber filtration funnel,
Filter Funnel, 3-piece, volume 210 mL, diam. 70 mm
Expand
View Pricing
WHA1950217
Whatman
®
glass microfiber filtration funnel,
Replacement 210 mL Reservoir, volume 400 mL, diam. 70 mm
Expand
View Pricing
WHA1950027
Whatman
®
glass microfiber filtration funnel,
3-piece, capacity 400 mL, diam. 70 mm
Expand
View Pricing
WHA1950207
Whatman
®
glass microfiber filtration funnel,
capacity 115 mL , reservoir, diam. 70 mm
Expand
View Pricing