Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Phloroglucinol
Synonym(s):
1,3,5-Trihydroxybenzene, Phloroglucinol
Linear Formula:
C
6
H
3
-1,3,5-(OH)
3
CAS Number:
108-73-6
Molecular Weight:
126.11
EC Number:
203-611-2
Products
Product Category
building blocks (1)
stains (1)
Brand
Sigma-Aldrich (4)
Supelco (1)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
79330
Phloroglucinol,
≥99.0% (HPLC)
Expand
View Pricing
1.07069
Phloroglucinol,
(1,3,5-trihydroxybenzene) for analysis
Expand
View Pricing
8.18887
Phloroglucinol,
for synthesis
Expand
View Pricing
P1178
Phloroglucinol,
suitable for plant cell culture, BioReagent
Expand
View Pricing
P3502
Phloroglucinol,
Used to detect the presence of wood fiber.
Expand
View Pricing