![Methyl chloroacetate 99%](/deepweb/assets/sigmaaldrich/product/structures/111/969/d49ba77c-437f-4cf7-8772-01ec662a6883/640/d49ba77c-437f-4cf7-8772-01ec662a6883.png)
Methyl chloroacetate
![Methyl chloroacetate 99%](/deepweb/assets/sigmaaldrich/product/structures/111/969/d49ba77c-437f-4cf7-8772-01ec662a6883/640/d49ba77c-437f-4cf7-8772-01ec662a6883.png)
Synonym(s):
Methyl chloroacetate
Linear Formula:
ClCH2COOCH3
CAS Number:
Molecular Weight:
108.52
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing