Skip to Content
Merck
All Photos(1)

Key Documents

8.02413

Sigma-Aldrich

Methyl chloroacetate

for synthesis

Synonym(s):

Methyl chloroacetate, Chloroacetic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5ClO2
CAS Number:
Molecular Weight:
108.52
MDL number:
UNSPSC Code:
12352108
EC Index Number:
202-501-1
NACRES:
NA.22
form:
liquid
Assay:
≥99.0% (GC)
Pricing and availability is not currently available.

vapor pressure

6.9 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

465 °C

potency

107 mg/kg LD50, oral (Rat)

expl. lim.

4.6-18.5 % (v/v)

pH

4.6 (20 °C, 28 g/L in H2O)

bp

130-132 °C/1013 hPa

mp

-33 °C

transition temp

flash point 52 °C (DIN 51758)

solubility

51.6 g/L (slow decomposition)

density

1.24 g/cm3 at 20 °C

SMILES string

ClCC(=O)OC

InChI

1S/C3H5ClO2/c1-6-3(5)2-4/h2H2,1H3

InChI key

QABLOFMHHSOFRJ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.234 - 1.236
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service