
Imidocarb dipropionate

Synonym(s):
N,N′-Bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea dipropanoate
Empirical Formula (Hill Notation):
C25H32N6O5
Products
Product Category
Formula Weight
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing