Skip to Content
Merck
All Photos(2)

Documents

33441

Supelco

Imidocarb dipropionate

VETRANAL®, analytical standard

Synonym(s):

N,N′-Bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea dipropanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H32N6O5
CAS Number:
Molecular Weight:
496.56
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

205 °C (dec.) (lit.)

application(s)

clinical testing

format

neat

SMILES string

CCC(O)=O.CCC(O)=O.O=C(Nc1cccc(c1)C2=NCCN2)Nc3cccc(c3)C4=NCCN4

InChI

1S/C19H20N6O.2C3H6O2/c26-19(24-15-5-1-3-13(11-15)17-20-7-8-21-17)25-16-6-2-4-14(12-16)18-22-9-10-23-18;2*1-2-3(4)5/h1-6,11-12H,7-10H2,(H,20,21)(H,22,23)(H2,24,25,26);2*2H2,1H3,(H,4,5)

InChI key

AFGQXWSHYUHHNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Imidocarb dipropionate is an antibiotic and diaminide of the carbanilide series of antiprotozoal compounds. It is commonly used in animals like horses, where it is found to be effective in inhibiting the Theileria equi strains causing infections.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Human-Animal Medicine - E-Book: Clinical Approaches to Zoonoses, Toxicants and Other Shared Health Risks (2009)
Robinson's Current Therapy in Equine Medicine - E-Book (2014)
Asier Galán et al.
PloS one, 13(1), e0190474-e0190474 (2018-01-06)
Canine babesiosis caused by the intraerythrocytic protozoan parasite Babesia canis is a tick-borne disease characterized by a host response that involves both cellular and humoral immunity. This study focuses on the secretion of cytokines Granulocyte-Macrophage Colony-Stimulating Factor (GM-CSF), Keratinocyte Chemotactic-like
Josipa Kuleš et al.
BMC veterinary research, 13(1), 82-82 (2017-04-02)
Canine babesiosis is a tick-borne disease caused by hemoprotozoan parasites of the genus Babesia. The disease can be clinically classified into uncomplicated and complicated forms. The aim of this study was to assess the level of endothelial activation and alterations
Gerhard Bringmann et al.
Veterinary parasitology, 283, 109177-109177 (2020-07-07)
Babesia canis is the predominant and clinically relevant canine Babesia species in Europe. Transmitted by vector ticks, the parasite enters red blood cells and induces a severe, potentially fatal hemolytic anemia. Here, we report on the antibabesial activities of three

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service