
Fenoxanil

Synonym(s):
N-(1-Cyano-1,2-dimethylpropyl)-2-(2,4-dichlorophenoxy)propionamide
Empirical Formula (Hill Notation):
C15H18Cl2N2O2
CAS Number:
Molecular Weight:
329.22
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1