Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Diamond shaped weigh boats
Products
Product Category
dishes (9)
Brand
Sigma-Aldrich (2)
Color
Manufacturer
Diameter (mm)
Height (mm)
Length (mm)
Volume (ml)
Package Size
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
HS1425AA
Diamond shaped weigh boats,
polystyrene, volume 5 mL, white, anti-static
Expand
View Pricing
W3887
Diamond shaped weigh boats,
polystyrene, volume 25 mL, white
Expand
View Pricing
HS1426AA
Diamond shaped weigh boats,
polystyrene, volume 5 mL, black, anti-static
Expand
View Pricing
W4137
Diamond shaped weigh boats,
polystyrene, volume 100 mL, white
Expand
View Pricing
HS1425A
Diamond shaped weigh boats,
polystyrene, volume 25 mL, white, anti-static
Expand
View Pricing
W4012
Diamond shaped weigh boats,
polystyrene, volume 30 mL, white
Expand
View Pricing
HS1425C
Diamond shaped weigh boats,
polystyrene, volume 100 mL, white, anti-static
Expand
View Pricing
HS1424AA
Diamond shaped weigh boats,
polystyrene, volume 5 mL, white
Expand
View Pricing
HS1425B
Diamond shaped weigh boats,
polystyrene, volume 30 mL, white, anti-static
Expand
View Pricing
HS1427C
Diamond shaped weigh boats,
polystyrene, volume 100 mL, black
Expand
View Pricing
HS1426B
Diamond shaped weigh boats,
polystyrene, volume 30 mL, black, anti-static
Expand
View Pricing
HS1426A
Diamond shaped weigh boats,
polystyrene, volume 25 mL, black, anti-static
Expand
View Pricing
HS1427AA
Diamond shaped weigh boats,
polystyrene, volume 5 mL, black
Expand
View Pricing
HS1426C
Diamond shaped weigh boats,
polystyrene, volume 100 mL, black, anti-static
Expand
View Pricing
HS1427B
Diamond shaped weigh boats,
polystyrene, volume 30 mL, black
Expand
View Pricing
HS1427A
Diamond shaped weigh boats,
polystyrene, volume 25 mL, black
Expand
View Pricing