Skip to Content
Merck
Brilliant Cresyl Blue ALD Certified by the Biological Stain Commission

Brilliant Cresyl Blue ALD

Brilliant Cresyl Blue ALD Certified by the Biological Stain Commission
Synonym(s):
Brilliant Blue C, Brilliant Cresyl blue
Linear Formula:
C17H20N3OCl · 1/2ZnCl2
CAS Number:
Molecular Weight:
385.96
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Application
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing