Skip to Content
Products
Cart
0
AE
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
BRAND
®
BLAUBRAND
®
measuring cylinder
Products
Product Category
cylinders (9)
Brand
Sigma-Aldrich (1)
Manufacturer
Height (mm)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z324353
BRAND
®
BLAUBRAND
®
measuring cylinder,
volume 100 mL, subdivision, 1 mL
Expand
View Pricing
Z324345
BRAND
®
BLAUBRAND
®
measuring cylinder,
volume 50 mL, subdivision, 1 mL
Expand
View Pricing
Z324329
BRAND
®
BLAUBRAND
®
measuring cylinder,
volume 10 mL
Expand
View Pricing
Z324361
BRAND
®
BLAUBRAND
®
measuring cylinder,
volume 250 mL
Expand
View Pricing
Z324396
BRAND
®
BLAUBRAND
®
measuring cylinder,
volume 1000 mL, subdivision, 10 mL
Expand
View Pricing
Z324388
BRAND
®
BLAUBRAND
®
measuring cylinder,
volume 500 mL, subdivision, 5 mL
Expand
View Pricing
Z324337
BRAND
®
BLAUBRAND
®
measuring cylinder,
volume 25 mL, subdivision, 0.5 mL
Expand
View Pricing
Z324310
BRAND
®
BLAUBRAND
®
measuring cylinder,
volume 5 mL
Expand
View Pricing
Z324418
BRAND
®
BLAUBRAND
®
measuring cylinder,
volume 2000 mL, subdivision, 20 mL
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Learn More