Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Assembled screw cap with hole with PTFE/silicone septum
Products
Product Category
septa (10)
Brand
Supelco (8)
Closure Type
Color
Package Size
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
27019
Assembled screw cap with hole with PTFE/silicone septum,
black polypropylene hole cap, thread for 15-425, PTFE/silicone, septum thickness 1.5 mm, for use with 7 mL vial, pkg of 100 ea
Expand
View Pricing
SU860078
Assembled screw cap with hole with PTFE/silicone septum,
black polypropylene hole cap, thread for 13-425, red PTFE/silicone, septum thickness 1.5 mm, for use with 4 mL vial, pkg of 100 ea
Expand
View Pricing
27022
Assembled screw cap with hole with PTFE/silicone septum,
black polypropylene cap, thread for 24-400, PTFE/silicone, septum thickness 3 mm, for use with 20, 40, or 60 mL vial, pkg of 100 ea
Expand
View Pricing
27273
Assembled screw cap with hole with PTFE/silicone septum,
black polypropylene hole cap, thread for 10-425, PTFE/silicone, septum thickness 1.5 mm, for use with 2 mL vial (large opening), pkg of 100 ea
Expand
View Pricing
27262
Assembled screw cap with hole with PTFE/silicone septum,
black polypropylene hole cap, thread for 8-425, PTFE/silicone, septum thickness 1.5 mm, for use with 2 mL vial (standard opening), pkg of 100 ea
Expand
View Pricing
27018
Assembled screw cap with hole with PTFE/silicone septum,
black polypropylene hole cap, thread for 13-425, PTFE/silicone, septum thickness 1.5 mm, for use with 4 mL vial, pkg of 100 ea
Expand
View Pricing
27261-U
Assembled screw cap with hole with PTFE/silicone septum,
black polypropylene hole cap, thread for 8-425, PTFE/silicone, Top Hat (septa and closure designed to fit together tightly), for use with 2 mL vial, 4.6 mm opening, pkg of 100 ea
Expand
View Pricing
SU860076
Assembled screw cap with hole with PTFE/silicone septum,
black polypropylene hole cap, thread for 8-425, PTFE/silicone (red PTFE/white silicone), septum thickness 1.3 mm, for use with 2 mL vial (standard opening), pkg of 100 ea
Expand
View Pricing
27020
Assembled screw cap with hole with PTFE/silicone septum,
black polypropylene hole cap, thread for 18-400, PTFE/silicone, septum thickness 1.5 mm, for use with 15 mL vial, pkg of 100 ea
Expand
View Pricing
854985
Assembled screw cap with hole with PTFE/silicone septum,
black polypropylene hole cap, thread for 8-425, PTFE/silicone (red PTFE/creme silicone), septum thickness 1.5 mm, for use with 2 mL vial (standard opening), pkg of 1000 ea
Expand
View Pricing