Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ammonium citrate dibasic
Synonym(s):
Ammonium hydrogencitrate, Citric acid ammonium salt, Diammonium hydrogen citrate
Linear Formula:
HOC(CO
2
H)(CH
2
CO
2
NH
4
)
2
CAS Number:
3012-65-5
Molecular Weight:
226.18
EC Number:
221-146-3
Products
Product Category
building blocks (2)
buffers (1)
Brand
Sigma-Aldrich (1)
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
ACS reagent (1)
Puriss (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
09833
Ammonium citrate dibasic,
BioXtra, ≥99.0% (T)
Expand
View Pricing
247561
Ammonium citrate dibasic,
ACS reagent, 98%
Expand
View Pricing
25102
Ammonium citrate dibasic,
puriss., ≥98%
Expand
View Pricing