Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Adapter for Eppendorf
®
S-4-104
Products
Product Category
adapters (13)
Manufacturer
Diameter (mm)
Height (mm)
Package Size
Technique
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
EP5825741005
Adapter for Eppendorf
®
S-4-104,
capacity, 250 mL flat, 175–250 mL, pack of 2
Expand
View Pricing
EP5825745000
Adapter for Eppendorf
®
S-4-104,
capacity, 1 x 500 mL Corning
®
tubes, pack of 2
Expand
View Pricing
EP5825733002
Adapter for Eppendorf
®
S-4-104,
capacity, 7 x 50 mL conical tubes, pack of 2
Expand
View Pricing
EP5825740009
Adapter for Eppendorf
®
S-4-104,
capacity, 1.5/2.0 mL, pack of 2
Expand
View Pricing
EP5825732006
Adapter for Eppendorf
®
S-4-104,
capacity, 5 x 50 mL conical tubes, pack of 2
Expand
View Pricing
EP5825744004
Adapter for Eppendorf
®
S-4-104,
capacity, 1 x wide neck bottle 750 mL, pack of 2
Expand
View Pricing
EP5825736001
Adapter for Eppendorf
®
S-4-104,
capacity, 20 x 5.5 - 12 mL round-bottom Tubes, pack of 2
Expand
View Pricing
EP5825747003
Adapter for Eppendorf
®
S-4-104,
capacity, 27 x 5 mL FACS, pack of 2
Expand
View Pricing
EP5825743008
Adapter for Eppendorf
®
S-4-104,
capacity, 20 x 9 mL round-bottom tubes, pack of 2
Expand
View Pricing
EP5920709003
Adapter for Eppendorf
®
S-4-104,
capacity, 10 x 50 mL conical tubes, for use with medium-capacity buckets, pack of 2
Expand
View Pricing
EP5825748000
Adapter for Eppendorf
®
S-4-104,
Holds 14 mL tubes, pack of 2
Expand
View Pricing
EP5825738004
Adapter for Eppendorf
®
S-4-104,
capacity, 23 x 2.6 - 8 mL round-bottom Tubes, pack of 2
Expand
View Pricing
EP5825755006
Adapter for Eppendorf
®
S-4-104,
Holds 30 mL tubes, pack of 2
Expand
View Pricing