Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Naphthol
Synonym(s):
β-Naphthol, 2-Hydroxynaphthalene, 2-Naphthol
Linear Formula:
C
10
H
7
OH
CAS Number:
135-19-3
Molecular Weight:
144.17
EC Number:
205-182-7
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (4)
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
185507
2-Naphthol,
99%
Expand
View Pricing
130109
2-Naphthol,
98%
Expand
View Pricing
8.22290
2-Naphthol,
for synthesis
Expand
View Pricing
70448
2-Naphthol,
BioXtra, ≥99.0% (GC)
Expand
View Pricing