Skip to Content
Merck
All Photos(1)

Key Documents

1663008

USP

Thioridazine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

10-[2-(1-Methyl-2-piperidyl)ethyl]-2-(methylthio)-10H-phenothiazine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2S2 · HCl
CAS Number:
Molecular Weight:
407.04
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

thioridazine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CSc1ccc2Sc3ccccc3N(CCC4CCCCN4C)c2c1

InChI

1S/C21H26N2S2.ClH/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)25-21-11-10-17(24-2)15-19(21)23;/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3;1H

InChI key

NZFNXWQNBYZDAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Thioridazine hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Thioridazine Hydrochloride Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service