Skip to Content
Merck
All Photos(1)

Key Documents

1598405

USP

Ranitidine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22N4O3S · HCl
CAS Number:
Molecular Weight:
350.86
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ranitidine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].CN\C(NCCSCc1ccc(CN(C)C)o1)=C\[N+]([O-])=O

InChI

1S/C13H22N4O3S.ClH/c1-14-13(9-17(18)19)15-6-7-21-10-12-5-4-11(20-12)8-16(2)3;/h4-5,9,14-15H,6-8,10H2,1-3H3;1H/b13-9-;

InChI key

GGWBHVILAJZWKJ-CHHCPSLASA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ranitidine hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Ranitidine Injection
  • Ranitidine Tablets
  • Ranitidine in Sodium Chloride Injection
  • Ranitidine Oral Solution

Biochem/physiol Actions

H2 histamine receptor antagonist; anti-ulcer agent.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Travis Nemkov et al.
JCI insight, 6(3) (2020-12-23)
Computational models based on recent maps of the RBC proteome suggest that mature erythrocytes may harbor targets for common drugs. This prediction is relevant to RBC storage in the blood bank, in which the impact of small molecule drugs or
Question 1: does the use of ranitidine increase the risk of NEC in preterm infants?
Manigandan Chandrasekaran et al.
Archives of disease in childhood, 99(4), 390-392 (2014-03-15)
M B Evans et al.
Journal of pharmaceutical and biomedical analysis, 7(1), 1-22 (1989-01-01)
The selection, development, definition and validation of selective stability-indicating procedures for high-performance liquid chromatographic and thin-layer chromatographic analyses of ranitidine hydrochloride are described. The procedures used in conjunction can be applied to the quality assurance and stability assessments of both
[The development of hemorrhagic vasculitis (Schönlein-Henoch disease) in a patient with a chronic stomach ulcer against a background of Sostril (ranitidine hydrochloride) therapy].
E N Glazunov et al.
Vestnik khirurgii imeni I. I. Grekova, 152(1-2), 102-103 (1994-01-01)
Natalia Alonso et al.
The Biochemical journal, 459(1), 117-126 (2014-01-15)
7TMRs (seven-transmembrane receptors) exist as conformational collections in which different conformations would lead to differential downstream behaviours such as receptor phosphorylation, G-protein activation and receptor internalization. In this context, a ligand may cause differential activation of some, but not all

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service