Skip to Content
Merck

1576800

USP

Propyl gallate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3,4,5-Trihydroxybenzoic acid propyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
3,4,5-(HO)3C6H2CO2CH2CH2CH3
CAS Number:
Molecular Weight:
212.20
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1877976
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

pharmaceutical primary standard

API family

propyl gallate

manufacturer/tradename

USP

mp

146-149 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCOC(=O)c1cc(O)c(O)c(O)c1

InChI

1S/C10H12O5/c1-2-3-15-10(14)6-4-7(11)9(13)8(12)5-6/h4-5,11-13H,2-3H2,1H3

InChI key

ZTHYODDOHIVTJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Preparation Note

Prepare a 0.1M solution in glycerol:PBS (9:1)

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

flash_point_f

368.6 °F - closed cup

flash_point_c

187 °C - closed cup

Storage Class

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheng-Kun Du et al.
Life sciences, 109(1), 15-19 (2014-06-17)
We have previously demonstrated that propyl gallate has a Ca(2+) sensitizing effect on the force generation in membrane-permeabilized (skinned) cardiac muscle fibers. However, in vivo beneficial effects of propyl gallate as a novel Ca(2+) sensitizer remain uncertain. In the present
Liliana O Rocha et al.
PloS one, 9(4), e92189-e92189 (2014-04-18)
Bacterial antagonists used as biocontrol agents represent part of an integrated management program to reduce pesticides in the environment. Bacillus thuringiensis is considered a good alternative as a biocontrol agent for suppressing plant pathogens such as Fusarium. In this study
International journal of toxicology, 26 Suppl 3, 89-118 (2008-02-19)
Propyl Gallate is the n-propyl ester of gallic acid (3,4,5-trihydroxybenzoic acid). It is soluble in ethanol, ethyl ether, oil, lard, and aqueous solutions of polyethylene glycol (PEG) ethers of cetyl alcohol, but only slightly soluble in water. Propyl Gallate currently
Amanda Psyrri et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(11), 3023-3032 (2014-04-05)
We sought to evaluate the correlation between tissue biomarker expression (using standardized, quantitative immunofluorescence) and clinical outcome in the E2303 trial. Sixty-three eligible patients with operable stage III/IV head and neck squamous cell cancer (HNSCC) participated in the Eastern Cooperative
Effects of dietary fat, butylated hydroxytoluene and propyl gallate on microsomal drug metabolism in rats.
Nutrition reviews, 40(6), 189-191 (1982-06-01)

Global Trade Item Number

SKUGTIN
1576800-200MG04061838743503

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service