Skip to Content
Merck
All Photos(1)

Key Documents

1576005

USP

Propranolol hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(±)-Propranolol hydrochloride, (±)-1-Isopropylamino-3-(1-naphthyloxy)-2-propanol hydrochloride, DL-Propranolol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H21NO2 · HCl
CAS Number:
Molecular Weight:
295.80
Beilstein:
4164259
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

propranolol

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.CC(C)NCC(O)COc1cccc2ccccc12

InChI

1S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18H,10-11H2,1-2H3;1H

InChI key

ZMRUPTIKESYGQW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Canan Akyüz et al.
Pediatric blood & cancer, 61(5), 931-932 (2013-11-23)
Lymphangiomas of the tongue are rare, and their treatment is problematic. A 10 year-old patient with tongue lymphangioma who was previously treated with surgery and propranolol with no response was treated with sirolimus in our department. We used sirolimus with
Katherine B Püttgen
Pediatric clinics of North America, 61(2), 383-402 (2014-03-19)
Propranolol has replaced corticosteroids as preferred first-line therapy for the management of infantile hemangiomas (IH). The topical β-blocker timolol is now an alternative to oral propranolol and watchful waiting for smaller IH. Research in the last decade has provided evidence-based
Ya-Yu Wang et al.
Endocrinology, 155(4), 1235-1246 (2014-01-21)
Patients who experience acute ischemic stroke may develop hyperglycemia, even in the absence of diabetes. In the current study we determined the effects of acute stroke on hepatic insulin signaling, TNF-α expression, endoplasmic reticulum (ER) stress, the activities of c-Jun
Pawel Szychta et al.
Plastic and reconstructive surgery, 133(4), 852-862 (2013-12-20)
Infantile hemangioma is a vascular tumor and requires treatment in lesions manifested by potentially dangerous symptoms. Several publications have reported that involution of infantile hemangioma could be accelerated by propranolol but have used only invalidated subjective measures of assessment. The
Dennis D Rasmussen et al.
Alcoholism, clinical and experimental research, 38(6), 1532-1539 (2014-06-04)
Evidence suggests that activation of the noradrenergic system may contribute to alcohol drinking in animals and humans. Our previous studies demonstrated that blocking α1 -adrenergic receptors with the antagonist, prazosin, decreased alcohol drinking in rats under various conditions. As noradrenergic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service