Skip to Content
Merck
All Photos(1)

Key Documents

1491015

USP

Oxytetracycline hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

5-Hydroxytetracycline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O9 · HCl
CAS Number:
Molecular Weight:
496.89
Beilstein:
3853107
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

oxytetracycline

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

Cl.CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3C(=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)C(=O)c4c(O)cccc4[C@@]3(C)O

InChI

1S/C22H24N2O9.ClH/c1-21(32)7-5-4-6-8(25)9(7)15(26)10-12(21)17(28)13-14(24(2)3)16(27)11(20(23)31)19(30)22(13,33)18(10)29;/h4-6,12-14,17,25,27-29,32-33H,1-3H3,(H2,23,31);1H/t12-,13-,14+,17+,21-,22+;/m1./s1

InChI key

UBDNTYUBJLXUNN-IFLJXUKPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxytetracycline hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhenxing Chi et al.
PloS one, 9(7), e102334-e102334 (2014-07-16)
The study investigated the effect of oxytetracycline (OTC) on the anti-oxidative defense system, the structure (hemolysis rate and morphology) and function (ATP enzyme activity) of human red blood cells (hRBCs) to investigate the possible toxic mechanism of OTC to hRBCs.
Alireza Khadem et al.
The British journal of nutrition, 112(7), 1110-1118 (2014-09-03)
The non-antibiotic anti-inflammatory theory of antimicrobial growth promoters (AGP) predicts that alternatives can be selected by simple in vitro tests. In vitro, the known AGP oxytetracycline (OTC) and a Macleaya cordata extract (MCE) had an anti-inflammatory effect with a half-maximal
Cağri Akyol et al.
Environmental technology, 35(17-20), 2373-2379 (2014-08-26)
Possible adverse effects of a commonly used veterinary antibiotic, oxytetracycline (OTC), on acidogenic phase of anaerobic digestion of cattle manure along with optimum operating conditions were investigated. A standard veterinary practice of 50 ml OTC solution (20 mg/kg cattle weight)
Chaoqi Chen et al.
Environmental pollution (Barking, Essex : 1987), 193, 94-101 (2014-07-13)
Non-irrigated and wastewater-irrigated soils were collected from five wastewater irrigation areas in Beijing and Tianjin, China. The concentrations of sulfadiazine, sulfamethoxazole, oxytetracycline and chlortetracycline in the soils were determined. Abundances of antibiotic resistant bacteria and corresponding resistance genes were also
Andrew P Baumann et al.
Bone, 75, 55-61 (2015-02-17)
Ulnar and tibial cyclic compression in rats and mice have become the preferred animal models for investigating the effects of mechanical loading on bone modeling/remodeling. Unlike rodents, rabbits provide a larger bone volume and normally exhibit intracortical Haversian remodeling, which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service