Skip to Content
Merck
All Photos(1)

Key Documents

1420006

USP

Levodopa Related Compound B

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

DL-3-O-Methyldopa, 3-Methoxytyrosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO4
CAS Number:
Molecular Weight:
211.21
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

levodopa

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

NC(Cc1cc(c(cc1)O)OC)C(=O)O

InChI

1S/C10H13NO4/c1-15-9-5-6(2-3-8(9)12)4-7(11)10(13)14/h2-3,5,7,12H,4,11H2,1H3,(H,13,14)

InChI key

PFDUUKDQEHURQC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levodopa Related Compound B USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Carbidopa and Levodopa Orally Disintegrating Tablets
  • Levodopa
  • Carbidopa and Levodopa Tablets
  • Carbidopa and Levodopa Extended-Release Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Müller et al.
Naunyn-Schmiedeberg's archives of pharmacology, 383(6), 627-633 (2011-05-03)
Elevation of plasma total homocysteine concentrations were observed in levodopa/dopa decarboxylase inhibitor (DDI)-treated patients with Parkinson's disease (PD). Degradation of levodopa to 3-O-methyldopa via the enzyme catechol-O-methyltransferase (COMT) is a methyl group demanding reaction. It generates homocysteine from the methyl
Stephanie L Merhar et al.
Pediatrics, 116(5), 1226-1230 (2005-11-03)
Chronic bilirubin encephalopathy, characterized clinically by extrapyramidal movement abnormalities, vertical gaze abnormalities, and hearing loss, results from neuronal injury after marked hyperbilirubinemia in term and preterm infants. In premature infants, bilirubin staining of specific brain structures has been described at
Li-Hsuan Wang et al.
Phytotherapy research : PTR, 24(6), 852-858 (2009-11-27)
The impacts of caffeic acid (3,4-dihydroxycinnamic acid, CA) on the pharmacokinetics of levodopa (L-dopa) were studied in rabbits. A single dose of 5/1.25 mg kg(-1) L-dopa/carbidopa was administered alone or was co-administered with three different doses of caffeic acid (2.5
Yoritaka Onzawa et al.
Biological & pharmaceutical bulletin, 35(8), 1244-1248 (2012-08-07)
It has been well known that 3-O-methyldopa (3-OMD) is a metabolite of L-3,4-dihydroxyphenylalanine (L-DOPA) formed by catechol O-methyltransferase (COMT), and 3-OMD blood level often reaches higher than physiological level in Parkinson's disease (PD) patients receiving long term L-DOPA therapy. However
T Müller et al.
Journal of neural transmission (Vienna, Austria : 1996), 114(11), 1457-1462 (2007-06-15)
The interval of line tracing performance is more associated to basal ganglia function due to the dependence on bradykinesia and rigidity. The other component of this task, the precision of execution of complex movement sequences, is more related to attention.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service