Skip to Content
Merck
All Photos(1)

Key Documents

1359415

USP

Levetiracetam Racemic Mixture

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(RS)-α-Ethyl-2-oxo-1-pyrrolidineacetamide, 2(RS)-(2-Oxopyrrolidin-1-yl)butyramide, Etiracetam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N2O2
CAS Number:
Molecular Weight:
170.21
Beilstein:
1529106
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

levetiracetam

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCC(N1CCCC1=O)C(N)=O

InChI

1S/C8H14N2O2/c1-2-6(8(9)12)10-5-3-4-7(10)11/h6H,2-5H2,1H3,(H2,9,12)

InChI key

HPHUVLMMVZITSG-UHFFFAOYSA-N

General description

Levetiracetam racemic mixture also know as etiracetam is an stuctural analogue ethylated piracetam drug.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levetiracetam Racemic Mixture USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Levetiracetam

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Can zinc depletion play a role in LEV-induced hair loss? Considerations from a case study.
Rocco Salvatore Calabrò et al.
Epilepsy & behavior : E&B, 29(1), 254-255 (2013-08-14)
Aaron M Cook et al.
Neurocritical care, 19(2), 210-214 (2013-08-03)
Increased creatinine clearance and subsequent elevated antimicrobial clearance is evident in many traumatic brain injury (TBI) patients due to augmented renal clearance (ARC). Little is known about the effects of ARC on other renally-eliminated medications, such as the anti-epileptic drug
S J Sara
Psychopharmacology, 68(3), 235-241 (1980-01-01)
Etiracetam, a nonanaleptic drug related to the nootropic substance piracetam, was found to facilitate memory retrieval in rats in several experimental situations, when injected 30 min prior to retention testing. The drug was active when memory deficits were induced by
Adrian Budhram et al.
Archives of physical medicine and rehabilitation, 95(3), 588-590 (2013-09-26)
Postanoxic myoclonus is a rare manifestation after an anoxic event, with fewer than 150 cases reported in the literature. The condition is characterized by myoclonic jerks, which are worse on action than at rest, and postural lapses, ataxia, and dysarthria.
Wenxia Luo et al.
Bioanalysis, 5(15), 1843-1851 (2013-08-03)
A simple LC-MS/MS method was developed and validated for the quantification of levetiracetam (LEV, Keppra®), a broad-spectrum antiepileptic drug (AED) in rat dried blood spots (DBS). LEV was simply extracted with methanol spiked with adenosine (ADE) as IS before LC-MS/MS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service