Skip to Content
Merck
All Photos(1)

Key Documents

1338801

USP

Indapamide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-(4-Chloro-3-sulfamoylbenzamido)-2-methylindoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16ClN3O3S
CAS Number:
Molecular Weight:
365.83
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

indapamide

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC1Cc2ccccc2N1NC(=O)c3ccc(Cl)c(c3)S(N)(=O)=O

InChI

1S/C16H16ClN3O3S/c1-10-8-11-4-2-3-5-14(11)20(10)19-16(21)12-6-7-13(17)15(9-12)24(18,22)23/h2-7,9-10H,8H2,1H3,(H,19,21)(H2,18,22,23)

InChI key

NDDAHWYSQHTHNT-UHFFFAOYSA-N

Gene Information

human ... SLC12A3(6559)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Indapamide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Indapamide Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Lact. - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Damien et al.
Clinical pharmacokinetics, 37 Suppl 1, 13-19 (1999-09-24)
In accordance with international guidelines recommending the use of low doses of antihypertensive agents, a new formulation of indapamide--indapamide sustained release (SR)--has been developed. Indapamide has been used worldwide for many years as an immediate release (IR) formulation at a
Sheng Kang et al.
Clinical therapeutics, 26(2), 257-270 (2004-03-25)
A low-dose combination of perindopril and indapamide may effectively reduce blood pressure (BP) in hypertensive patients, but some factors related to study design might have contributed to the between-group differences in the rate of reduction of BP observed in some
Dean M Robinson et al.
Drugs, 66(2), 257-271 (2006-02-03)
A low-dose sustained-release (SR) formulation of the thiazide-type diuretic indapamide, indapamide SR (Natrilix SR), retains the antihypertensive activity of the immediate-release (IR) formulation, with a smoother pharmacokinetic profile. In well controlled 12- to 52-week clinical trials, indapamide SR 1.5 mg/day
A J Matheson et al.
Drugs, 61(8), 1211-1229 (2001-07-24)
The fixed low-dose combination of the ACE inhibitor perindopril and the non-thiazide diuretic indapamide has been evaluated in the management of patients with mild to moderate hypertension. Combination therapy aims to improve overall therapeutic efficacy while minimising adverse effects. In
J Sassard et al.
Fundamental & clinical pharmacology, 19(6), 637-645 (2005-11-30)
The relationship between blood pressure (BP) and cardiovascular risk is clearly established; hypertension increases the rate of cardiovascular. High systolic blood pressure (SBP) may be the main parameter involved in cardiovascular morbidity and mortality. The benefit of lowering BP, particularly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service