Skip to Content
Merck
All Photos(1)

Documents

1299007

USP

Griseofulvin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(2S)-trans-7-Chloro-2′,4,6-trimethoxy-6′-methylspiro(benzofuran-2[3H],1′-[2]cyclohexene)-3,4′-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17ClO6
CAS Number:
Molecular Weight:
352.77
Beilstein:
95226
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

griseofulvin

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

ClC1=C(O[C@@]2(C(OC)=CC(C[C@H]2C)=O)C3=O)C3=C(OC)C=C1OC

InChI

1S/C17H17ClO6/c1-8-5-9(19)6-12(23-4)17(8)16(20)13-10(21-2)7-11(22-3)14(18)15(13)24-17/h6-8H,5H2,1-4H3/t8-,17+/m1/s1

InChI key

DDUHZTYCFQRHIY-RBHXEPJQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Griseofulvin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Griseofulvin Capsules
  • Griseofulvin Tablets
  • Ultramicrosize Griseofulvin Tablets
  • Griseofulvin Oral Suspension

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Damir Elmar Zecevic et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 87(2), 264-270 (2014-04-29)
Many upcoming drug candidates are pH-dependent poorly soluble weak bases in the pH range of the gastrointestinal tract. This often leads to a high in vivo variability and bioavailability issues. Aiming to overcome these limitations, the design of solid dispersions
Mark Sacchetti
Journal of pharmaceutical sciences, 103(9), 2772-2783 (2013-12-12)
Pharmaceutical materials, crystalline and amorphous, sorb water from the atmosphere, which affects critical factors in the development of drugs, such as the selection of drug substance crystal form, compatibility with excipients, dosage form selection, packaging, and product shelf-life. It is
Ildikó Nyilasi et al.
Medical mycology, 52(2), 140-148 (2013-09-06)
The investigation of the antifungal activities of drugs whose primary activities are not related to their antimicrobial potential is in the current forefront of research. Statin compounds, which are routinely used as cholesterol-lowering drugs, may also exert direct antimicrobial effects.
Dipy M Vasa et al.
Journal of pharmaceutical sciences, 103(9), 2911-2923 (2014-05-16)
Fifteen model drugs were quenched from 3:1 (w/w) mixtures with polyethylene glycol 4000 (PEG4000). The resulting solids were characterized using powder X-ray diffraction (PXRD), analysis of pair distribution function-transformed PXRD data (where appropriate), hot-stage polarized light microscopy, and differential scanning
Kayo Yuminoki et al.
Journal of pharmaceutical sciences, 103(11), 3772-3781 (2014-09-12)
In this study, we reported the application of Povacoat®, a hydrophilic polyvinylalcohol copolymer, as a dispersion stabilizer of nanoparticles of poorly water-soluble compounds. In addition, the influence of aggregation of the nanoparticles on their solubility and oral absorption was studied.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service