Skip to Content
Merck
All Photos(1)

Documents

1211006

USP

Dimethyl sulfoxide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

DMSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
Beilstein:
506008
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

manufacturer/tradename

USP

availability

not available in (Sales restrictions may apply)

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)

mp

16-19 °C (lit.)

density

1.10 g/mL (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dimethyl sulfoxide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Dimethyl Sulfoxide Irrigation
  • Dimethyl Sulfoxide Gel
  • Dimethyl Sulfoxide Topical Solution
  • Methylsulfonylmethane

Caution

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methylsulfonylmethane
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 32(3), 5097-5097 (2020)
Dimethyl Sulfoxide Topical Solution
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 1414-1414 (2020)
Björn Zörner et al.
Brain : a journal of neurology, 137(Pt 6), 1716-1732 (2014-04-17)
Anatomical plasticity such as fibre growth and the formation of new connections in the cortex and spinal cord is one known mechanism mediating functional recovery after damage to the central nervous system. Little is known about anatomical plasticity in the
Mulmudi Hemant Kumar et al.
Advanced materials (Deerfield Beach, Fla.), 26(41), 7122-7127 (2014-09-13)
Lead free perovskite solar cells based on a CsSnI3 light absorber with a spectral response from 950 nm is demonstrated. The high photocurrents noted in the system are a consequence of SnF2 addition which reduces defect concentrations and hence the
Phillip McDonagh et al.
Veterinary microbiology, 174(3-4), 438-447 (2014-12-04)
Feline infectious peritonitis (FIP), a feline coronavirus (FCoV) induced disease, is almost invariably fatal with median life expectancy measured in days. Current treatment options are, at best, palliative. The objectives of this study were to evaluate a panel of nineteen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service