Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C15H10O4
CAS Number:
Molecular Weight:
254.24
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
231523
Pricing and availability is not currently available.
grade
pharmaceutical primary standard
agency
USP
API family
daidzein
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
−20°C
SMILES string
Oc1ccc(cc1)C2=COc3cc(O)ccc3C2=O
InChI
1S/C15H10O4/c16-10-3-1-9(2-4-10)13-8-19-14-7-11(17)5-6-12(14)15(13)18/h1-8,16-17H
InChI key
ZQSIJRDFPHDXIC-UHFFFAOYSA-N
General description
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Daidzein USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
- Red Clover Tablets
- Red Clover Aerial Parts Isoflavone Aglycones Dry Extract
- Powdered Red Clover
- Red Clover
- Soy Isoflavones Capsules
- Soy Isoflavones Tablets
- Powdered Red Clover Extract
Biochem/physiol Actions
Soy isoflavone daidzein protects against oxidative damage in liver cells induced by 7,12-dimethylbenz[a]anthracene (DMBA). Catalase and superoxide dismutase activity, down-regulated by DMBA, was restored by daidzein.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename - | manufacturer/tradename Calbiochem® |
| grade pharmaceutical primary standard | grade certified reference material, pharmaceutical secondary standard | grade analytical standard | grade - |
| format neat | format - | format neat | format - |
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical | application(s) food and beverages | application(s) - |
| API family daidzein | API family daidzein | API family - | API family - |
| storage temp. −20°C | storage temp. -10 to -25°C | storage temp. 2-8°C | storage temp. −20°C |
Still not finding the right product?
Explore all of our products under Daidzein
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


