Skip to Content
Merck
All Photos(1)

Key Documents

1142108

USP

Clozapine Resolution Mixture

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Clozapine, 8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]-diazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H19ClN4
CAS Number:
Molecular Weight:
326.82
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clozapine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1CCN(CC1)C2=Nc3cc(Cl)ccc3Nc4ccccc24

InChI

1S/C18H19ClN4/c1-22-8-10-23(11-9-22)18-14-4-2-3-5-15(14)20-16-7-6-13(19)12-17(16)21-18/h2-7,12,20H,8-11H2,1H3

InChI key

QZUDBNBUXVUHMW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clozapine Resolution Mixture USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Clozapine
  • Clozapine Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saeko Ikai et al.
The Annals of pharmacotherapy, 47(7-8), e31-e31 (2013-06-13)
To report on a patient who was successfully rechallenged with clozapine after perforation of the large intestine and pulmonary embolism post operatively, and provide a literature review on clozapine rechallenge. A 46-year-old Japanese man with treatment-resistant schizophrenia developed constipation and
Benjamin W J Spencer et al.
Pharmacogenomics, 14(15), 1907-1914 (2013-11-19)
Clozapine is a uniquely efficacious antipsychotic drug in treatment-resistant schizophrenia. Its use is restricted due to adverse effects including a rare but dangerous reduction in neutrophils (agranulocytosis) and the mandatory hematological monitoring this entails in many countries. We review the
Rasmus Røge et al.
Schizophrenia research, 140(1-3), 204-213 (2012-07-27)
Clozapine remains the drug of choice for treatment resistant schizophrenia, but is associated with potentially life threatening side effects, including agranulocytosis and myocarditis. Immunological mechanisms may be involved in the development of these side effects or in the unique antipsychotic
A Merenlender-Wagner et al.
Molecular psychiatry, 20(1), 126-132 (2013-12-25)
Autophagy is a process preserving the balance between synthesis, degradation and recycling of cellular components and is therefore essential for neuronal survival and function. Several key proteins govern the autophagy pathway including beclin1 and microtubule associated protein 1 light chain
Gary Remington et al.
Psychopharmacology, 225(3), 505-518 (2012-11-28)
Clozapine levels are advocated in the monitoring of patients on this drug and have now been used for a number of years. A safety-related threshold has also been proposed, as well as therapeutic lower and upper thresholds. While there has

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service