Skip to Content
Merck
All Photos(1)

Documents

1004001

USP

Acetanilide Melting Point Standard

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Acetanilide, N-Phenylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H5
CAS Number:
Molecular Weight:
135.16
Beilstein:
606468
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

4.65 (vs air)

vapor pressure

1 mmHg ( 114 °C)

API family

acetanilide

autoignition temp.

1004 °F

manufacturer/tradename

USP

bp

304 °C (lit.)

mp

113-115 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(=O)Nc1ccccc1

InChI

1S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10)

InChI key

FZERHIULMFGESH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

For further information and support please go to the website of the issuing Pharmacopoeia.

Application

This USP reference standard is intended for use in specified quality tests and assays.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

321.8 °F - closed cup

Flash Point(C)

161 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Edler et al.
Physical review letters, 88(6), 067403-067403 (2002-02-28)
Femtosecond IR spectroscopy of delocalized NH excitations of crystalline acetanilide confirms that self-trapping in hydrogen-bonded peptide units exists and does stabilize the excitation. Two phonons with frequencies of 48 and 76 cm (-1) are identified as the major degrees of
Fabrice Gritti et al.
Journal of chromatography. A, 1355, 179-192 (2014-07-09)
The mass transfer mechanism in four prototype columns (2.1 and 3.0×50mm, 2.1 and 3.0×100mm) packed with 1.9μm fully porous Titan-C18 particles was investigated by using two previously reported home-made protocols. The first one was used to measure the eddy dispersion
Naoki Aizawa et al.
Neurourology and urodynamics, 34(4), 368-374 (2014-02-18)
We measured single-unit mechanosensitive afferent activities (SAAs) during reflexic, rhythmic bladder contractions (RBCs), and examined whether L-arginine, an NO substrate, and mirabegron, a β3-adrenoceptor agonist, and oxybutynin, an antimuscarinic agent, can affect the SAAs in such condition. Twenty-nine female Sprague-Dawley
Hassan A Alhazmi et al.
Journal of pharmaceutical and biomedical analysis, 107, 311-317 (2015-02-02)
In this work, the behavior of several metal ions with different globular proteins was investigated by affinity capillary electrophoresis. Screening was conducted by applying a proper rinsing protocol developed by our group. The use of 0.1M EDTA in the rinsing
Thomas Hahne et al.
Journal of bioscience and bioengineering, 118(3), 311-314 (2014-04-04)
Human papillomavirus 6b L1 virus-like particles (VLPs) were successfully expressed using Bombyx mori nucleopolyhedrovirus (BmNPV) bacmid expression system and rapidly purified using size exclusion chromatography after ultracentrifugation procedure and characterized by capillary zone electrophoresis (CZE). The average capillary electrophoresis migration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service