Key Documents
RAD196
radiello Outdoor Shelter
pk of 10
About This Item
UNSPSC Code:
41104008
NACRES:
NB.24
packaging
pk of 10
manufacturer/tradename
Radiello Code 196
technique(s)
passive air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
General description
A polypropylene protective outdoor shelter is available for housing up to four Radiello samplers during outdoor/ambient air sampling. The shelter allows for adequate ventilation while simultaneously protecting the samplers from harsh weather conditions. The shelter can be mounted to a variety of fixtures including lamp posts, traffic lights, and telephone poles of various diameters. The shelter can easily be tranported from the lab and mounted without the use of tools. Two mounting strips are included with each shelter.
Legal Information
radiello is a trademark of Institi Clinici Scientifici Maugeri
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
J Dekeirsschieter et al.
Forensic science international, 189(1-3), 46-53 (2009-05-09)
Forensic entomology uses pig carcasses to surrogate human decomposition and to investigate the entomofaunal colonization. Insects communicate with their environment through the use of chemical mediators, which in the case of necrophagous insects, may consist in the cadaveric volatile organic
Benoît Oury et al.
Journal of occupational and environmental hygiene, 3(10), 547-557 (2006-08-16)
Diffusive sampling is particularly suited to determine time-weighted average volatile organic compound (VOC) concentration in occupational hygiene and environmental air monitoring. The purpose of this study was to measure the sampling rate variation of four different samplers in a special
Francesc A Esteve-Turrillas et al.
Journal of chromatography. A, 1216(48), 8549-8556 (2009-10-27)
A new procedure for the passive sampling in air of benzene, toluene, ethylbenzene and xylene isomers (BTEX) is proposed. A low-density polyethylene layflat tube filled with a mixture of solid phases provided a high versatility tool for the sampling of
P Bruno et al.
Waste management (New York, N.Y.), 27(4), 539-544 (2006-05-23)
Nuisance caused by odors is one of the most important problems for waste management plants. To control an odor nuisance, it must first be quantified. The analytical difficulties in odor measurements are related to the high number of volatile components
Alberto Sturaro et al.
Environmental monitoring and assessment, 164(1-4), 93-100 (2009-04-04)
This paper reports the results of a long-term monitoring of benzene, toluene, ethylbenzene, xylenes (BTEX), phenol and cresols in the air of Padua during a wide period of the year 2007 using two radial passive samplers (Radiello system) equipped with
Articles
Learn the how the diffusive air sampler works to see if it’s the right fit for you.
Learn the how the diffusive air sampler works to see if it’s the right fit for you.
Protocols
Learn how to assemble the supporting plate of your radiello air sampler in the lab before the sampling campaign to save time in the field.
Related Content
Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.
Learn about radiello diffusive sampling and why it’s the best option for your air quality needs.