Skip to Content
Merck
All Photos(1)

Documents

EURM019

Single stranded RNA (ssRNA) fragments of SARS-CoV-2

EURM Reference Material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

manufacturer/tradename

JRC

application(s)

clinical testing

storage temp.

−20°C

General description

Intended for the use as a positive quality control sample in the testing for the presence of the coronavirus SARS-CoV-2.
It can be used to verify the correctness of the transcription and amplification steps for the SARS-CoV-2 real-time RT-PCR assays.

EURM-019 is a solution containing a stabilised in vitro transcripted (IVT) synthetic single stranded RNA (ssRNA) in buffer. It does not contain any viable virus.

This universal synthetic ssRNA of 880 nt contains the target regions that can be amplified by the following RT-PCR assays:
a) N1, N2 and N3 gene developed by the Centers for Disease Control and Prevention (USA)
b) E gene and the RdRP gene developed by the Charité (DE)
c) N gene developed by the Japanese National Institute of Infectious Diseases (JP)
d) N gene developed by the Ministry of Heath of Thailand (TH)
e) S gene developed by the Joint Research Centre of the European Commission (EU)

Other Notes

for more information please see:
EURM019

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service