CRM78263
Ethylene glycol butyl ether
certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
2-Butoxyethanol, Butyl glycol
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
vapor density
4.1 (vs air)
vapor pressure
<1 mmHg ( 20 °C)
10 mmHg ( 81 °C)
product line
TraceCERT®
autoignition temp.
473 °F
shelf life
limited shelf life, expiry date on the label
expl. lim.
10.6 %
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
refractive index
n20/D 1.419 (lit.)
bp
169-172.5 °C (lit.)
mp
−75 °C (lit.)
density
0.902 g/mL at 25 °C (lit.)
storage temp.
−20°C
SMILES string
CCCCOCCO
InChI
1S/C6H14O2/c1-2-3-5-8-6-4-7/h7H,2-6H2,1H3
InChI key
POAOYUHQDCAZBD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point(F)
152.6 °F - Pensky-Martens closed cup
Flash Point(C)
67 °C - Pensky-Martens closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service