Skip to Content
Merck
All Photos(1)

Documents

CRM48743

Supelco

EPA 610 Polynuclear Aromatic Hydrocarbons Mix

certified reference material, in methanol: methylene chloride (1:1) (varied concentration)

Synonym(s):

EPA 610 Polynuclear Aromatic Hydrocarbons Mixture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 610
610

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

in methanol: methylene chloride (1:1) (varied concentration)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

We offer a variety of chromatography accessories including analytical syringes

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acenaphthene 1000 μg/mL
    Acenaphthylene 2000 μg/mL
    Anthracene 100 μg/mL
    Benz[a]anthracene 100 μg/mL
    Benzo[b]fluoranthene 200 μg/mL
    Benzo[k]fluoranthene 100 μg/mL
    Benzo[ghi]perylene 200 μg/mL
    Benzo[a]pyrene 100 μg/mL
    Chrysene 100 μg/mL
    Dibenz[ah]anthracene 200 μg/mL
    Fluoranthene 200 μg/mL
    Fluorene 200 μg/mL
    Indeno[1,2,3-cd]pyrene 100 μg/mL
    Naphthalene 1000 μg/mL
    Phenanthrene 100 μg/mL
    Pyrene 100 μg/mL
    See All (16)

Signal Word

Danger

Flash Point(F)

51.8 °F

Flash Point(C)

11.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Ascentis® Express PAH column efficiently separates polynuclear aromatic hydrocarbons in EPA methods.

Protocols

GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.

GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.

GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.

GCxGC excels in compound resolution, making it indispensable in analytical chemistry for complex samples.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service