Skip to Content
Merck
All Photos(1)

Key Documents

CRM44794

Supelco

Phosphamidon

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2-Chloro-N,N-diethyl-3-(dimethylphosphono)crotonic amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19ClNO5P
CAS Number:
Molecular Weight:
299.69
Beilstein:
8323501
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

−20°C

SMILES string

CCN(CC)C(=O)\C(Cl)=C(\C)OP(=O)(OC)OC

InChI

1S/C10H19ClNO5P/c1-6-12(7-2)10(13)9(11)8(3)17-18(14,15-4)16-5/h6-7H2,1-5H3/b9-8+

InChI key

RGCLLPNLLBQHPF-CMDGGOBGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Phosphamidon is a broad-spectrum systemic organophosphorus pesticide. It is used as an insecticide against sap-feeding insects, rice-stem borers, and rice leaf beetles in crops of sugarcane, citrus fruits, commercial cotton, and nuts.
As per Regulation (EC) No. 1107/2009, repealing directive 91/414/EEC, phosphamidon is not approved for use in the European Union. But a default maximum residue limit (MRL) of 0.01 mg/kg is applicable for the presence of phosphamidon in products of plant origin, in accordance with Regulation (EC) No. 149/2008.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
The phosphamidon CRM may also find following uses:
  • Determination of five organophosphorus pesticides in a medicinal herb sample using QuEChERS along with dispersive liquid-liquid microextraction (DLLME) and stacking followed by micellar electrokinetic chromatography (MEKC)
  • Magnetic solid phase microextraction (MSPE) of five organophosphorus pesticides using graphene based cyanopropyltriethoxysilane as an adsorbent from cow′s milk samples and their subsequent determination by gas chromatography with micro-electron capture detector
  • Investigation of residue levels of highly toxic pesticides in 6554 samples of fruits and vegetables by quadrupole time-of-flight LC/MS
  • Analysis of 4 organophosphorus pesticides by gas chromatography with microelectron capture detector following their extraction using a newly synthesized magnetic hybrid material as a solid phase extraction adsorbent
  • Multi-residue analysis of 44 organophosphorus pesticides by QuEChERS method combined with gas chromatography in Pogostemon cablin samples
  • Detection of organophosphorus pesticides in Chinese cabbage sample by a newly designed positive photoionization-ion mobility spectrometry (PP-IMS) apparatus

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Muta. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qinghua Zhou et al.
Talanta, 176, 247-252 (2017-09-18)
Ion mobility spectrometry (IMS) opened a potential avenue for the rapid detection of organophosphorus pesticides (OPPs), though an improved selectivity of stand-alone IMS was still in high demand. In this study, a stand-alone positive photoionization ion mobility spectrometry (PP-IMS) apparatus
Photocatalytic degradation of phosphamidon using Ag-doped ZnO nanorods
Korake PV, et al.
Toxicological & Environmental Chemistry, 94(6), 1075-1085 (2012)
Somayeh Koupaei Malek et al.
Journal of separation science (2018-06-08)
The precise control of pesticide residues in foodstuffs depends significantly on the clean extraction of analytes using specifically designed separation methods. In this study, a one-pot sol-gel process was used for the preparation of a magnetic hybrid silica gel tetraethylortho
Magnetic graphene-based cyanopropyltriethoxysilane as an adsorbent for simultaneous determination of polar and non-polar organophosphorus pesticides in cow?s milk
Nodeh H R, et al.
Royal Society of Chemistry Advances, 6(30), 24853-24864 (2016)
Trace determination of five organophosphorus pesticides by using QuEChERS coupled with dispersive liquid-liquid microextraction and stacking before micellar electrokinetic chromatography
Wei J, et al.
Analytical Methods : Advancing Methods and Applications, 7(14), 5801-5807 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service