Skip to Content
Merck
All Photos(1)

Key Documents

CRM40937

Supelco

Terpene Mix B

certified reference material, 2000 μg/mL each component in methanol, ampule of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in methanol

technique(s)

gas chromatography (GC): suitable

application(s)

cannabis testing
cannabis testing

format

multi-component solution

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Essential Oil Analysis Essential oils are complex mixtures of chemical compounds produced by living organisms and isolated from specific plant parts of known taxonomic origin. Their composition mainly includes monoterpenes, sesquiterpenes, and their oxygenated derivatives, as well as aldehydes, alcohols, and esters. In collaboration with Chromaleont and Shimadzu, our integrated solutions enhance efficiency and sustainability for the analysis of essential oils, addressing demanding workflows through innovative research, case studies, consumables, and applications.Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    beta-Pinene
    beta-Caryophyllene
    Phytol
    Limonene
    Geraniol
    (1S)-(-)-Camphor
    Terpinolene
    beta-Eudesmol
    (+)-Borneol
    cis-Nerolidol
    (-)-alpha-Terpineol
    (1S)-(+)-3-Carene
    Linalool
    p-Cymene
    See All (14)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Giuseppe Mannino et al.
International journal of molecular sciences, 22(5) (2021-04-04)
Hypercholesterolemia is one of the major causes of cardiovascular disease, the risk of which is further increased if other forms of dyslipidemia occur. Current therapeutic strategies include changes in lifestyle coupled with drug administration. Statins represent the most common therapeutic

Articles

Learn more about how the thermal desorption technique is applied for terpene identification, profiling, odor measurement, industrial hygiene, and environmental emissions.

Learn more about how the thermal desorption technique is applied for terpene identification, profiling, odor measurement, industrial hygiene, and environmental emissions.

Learn more about how the thermal desorption technique is applied for terpene identification, profiling, odor measurement, industrial hygiene, and environmental emissions.

Learn more about how the thermal desorption technique is applied for terpene identification, profiling, odor measurement, industrial hygiene, and environmental emissions.

See All

Protocols

Complete workflow for the comprehensive analysis of terpenes in cannabis

Complete workflow for the comprehensive analysis of terpenes in cannabis

Chromatograms

suitable for GC/MS, suitable for solid phase microextraction (SPME)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service