Skip to Content
Merck
All Photos(1)

Key Documents

CRM18124

Supelco

Mesotrion Metabolite AMBA

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

2-Amino-4-(methylsulfonyl)benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO4S
CAS Number:
Molecular Weight:
215.23
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

−20°C

SMILES string

[S](=O)(=O)(C)c1cc(c(cc1)C(=O)O)N

InChI

1S/C8H9NO4S/c1-14(12,13)5-2-3-6(8(10)11)7(9)4-5/h2-4H,9H2,1H3,(H,10,11)

InChI key

KFOGGDGNOLZBNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

AMBA (2-amino-4-methylsulfonylbenzoic acid) is one of the metabolites formed from biodegradation of parent Mesotrione. Mesotrione is a selective herbicide that belongs to the triketone family, developed for use in maize culture as a substitute for atrazine. It is used for pre- and post-emergence control of common broad-leaved weeds, primarily in corn (maize). It acts by inhibiting the p-hydroxyphenylpyruvate dioxygenase (HPPD) enzyme, thus, blocking the production of carotenoid pigments in susceptible plants.

Mesotrion is currently approved for its use in the European Union. Maximum residue levels (MRLs) have been set according to Reg (EU) 2017/626 for mesotrione for various products of plant and animal origin from 0.01 to 0.05 mg/kg. Compared to the parent compound, AMBA metabolite is more persistent in soil and possess greater toxicity in selected microorganisms.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The CRM may also find following uses:
  • Study the biodegradation and toxicity of herbicide mixtures mesotrione, nicosulfuron, and S-metolachlor by a bacterial strain, Bacillus sp. Mes11
  • Isolation and characterization of a Bacillus sp. from cloud water for rapid biotransformation of mesotrione herbicide
  • Investigation of mesotrione biodegradation products by the bacterial strain Bacillus sp. 3B6 using liquid chromatography/electrospray ionization quadrupole time-of-flight mass spectrometry (LC/ESI-QTOF-MS)
  • Qualitative and quantitative metabolic profiling of mesotrione, by the bacterial strain Bacillus sp. 3B6 using NMR, LC-NMR, and LC-MS
  • Method development for the determination of the mesotrione and its two metabolites, MNBA and AMBA in rice, rice plants, rice hulls, water, and soil using UPLC and triple-quadrupole tandem mass spectrometry

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kassio Ferreira Mendes et al.
Environmental science and pollution research international, 24(9), 8425-8435 (2017-02-12)
The effects of mesotrione, S-metolachlor, and terbuthylazine, applied in mixture, on soil biodegradation remain insufficiently researched. However, herbicide mixtures have been a common practice in agricultural systems in the last years. Understanding the fate of soil-applied herbicides may help on
Stéphanie Durand et al.
Rapid communications in mass spectrometry : RCM, 20(17), 2603-2613 (2006-08-01)
The metabolic pathway involved in the biotransformation of the herbicide mesotrione by the bacterial strain Bacillus sp. 3B6 was investigated by a reliable liquid chromatography/electrospray ionization quadrupole time-of-flight mass spectrometry (LC/ESI-QTOF-MS) method. The LC/ESI-MS method, both in positive and negative
Pengqiang Du et al.
Food chemistry, 229, 260-267 (2017-04-05)
In this study, an efficient and fast method for the determination of the herbicide mesotrione and its two metabolites [4-methylsulfonyl-2-nitrobenzoic acid (MNBA) and 2-amino-4-methylsulfonylbenzoic acid (AMBA)] in rice, rice plants, rice hulls, water, and soil was developed, using ultra performance
Isabelle Batisson et al.
Environmental pollution (Barking, Essex : 1987), 157(4), 1195-1201 (2009-01-06)
Dissipation kinetics of mesotrione, a new triketone herbicide, sprayed on soil from Limagne (Puy-de-Dôme, France) showed that the soil microflora were able to biotransform it. Bacteria from this soil were cultured in mineral salt solution supplemented with mesotrione as sole
S Durand et al.
Letters in applied microbiology, 43(2), 222-228 (2006-07-28)
The aim of this study was to find and characterize a fungal or bacterial strain capable of metabolizing mesotrione, a new selective herbicide for control of broad-leaved weeds in maize. This strain was isolated from cloud water and showed close

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service