Skip to Content
Merck
All Photos(1)

Documents

CRM05394

Supelco

Fenazaquin

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N2O
CAS Number:
Molecular Weight:
306.40
Beilstein:
8331263
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

CC(C)(C)c1ccc(CCOc2ncnc3ccccc23)cc1

InChI

1S/C20H22N2O/c1-20(2,3)16-10-8-15(9-11-16)12-13-23-19-17-6-4-5-7-18(17)21-14-22-19/h4-11,14H,12-13H2,1-3H3

InChI key

DMYHGDXADUDKCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Fenazaquin is an insecticide/acaricide that belongs to the class of quinazoline compounds. It shows ovicidal activity against phytophagus mites found in agricultural crops of almonds, citrus fruits, and grapes. It is highly lipophilic and inhibits site I of the electron transport chain in mitochondria. Its acceptable daily intake (ADI) according to body weight is 0.005 mg/kg.
Fenazaquin was approved for use in the European Union on 1st June 2011 following a request for its inclusion according to Reg. 33/2008 and its approval for use expires on 31st May 2023. The maximum residue limit (MRL) for fenazaquin in citrus fruits is 0.5 mg/kg and 0.2 mg/kg in tree nuts. Fenazaquin is to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
It may also find following uses:
  • Determination of trace levels of fenazaquin in raw almond samples using dispersive magnetic solid phase extraction (DMSPE) and gas chromatography-mass spectrometry (GC-MS)
  • Development of a liquid phase microextraction method using eutectic solvents for extraction from different fruit and vegetable samples and gas chromatography determination
  • Switchable solvent microextraction of pesticide residues from food and environmental samples
  • Evaluation of 60 pesticide residues in 144 different fruit and vegetable samples by LC-MS/MS method
  • Study of matrix effects in pesticide residue analysis in different citrus fruits using ethyl acetate for extraction and Ultrahigh-Performance Liquid Chromatography–High-Resolution Mass Spectrometry for determination
  • Residue analysis of 64 pesticides from various groups by nanoflow liquid chromatography-mass spectrometry (LC-MS)

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalia Besil et al.
Journal of agricultural and food chemistry, 65(23), 4819-4829 (2017-05-26)
The matrix effects of ethyl acetate extracts from seven different citrus fruits on the determination of 80 pesticide residues using liquid chromatography coupled to high-resolution time-of-flight mass spectrometry (UHPLC-(ESI)-HR-TOF) at 4 GHz resolution mode were studied. Only 20% of the
Herbert Musarurwa et al.
Talanta, 224, 121807-121807 (2021-01-01)
Switchable solvents are new generation solvents that are environmentally friendly and can be used for the effective pre-concentration of pesticide residues in food and environmental matrices. They have physico-chemical properties that can be switched abruptly and reversibly between two opposite
Anna Stachniuk et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 52(7), 446-457 (2017-03-30)
The aim of the research is to evaluate pesticide residue contamination of fresh and frozen fruits and vegetables, agricultural raw material, purchased from Polish farmers for production of frozen fruits and vegetables, and the estimation of the multiresidue method effectiveness
Application of oleic acid functionalized magnetic nanoparticles for a highly sensitive and efficient dispersive magnetic solid phase extraction of fenazaquin in almond samples for determination by gas chromatrography mass spectrometry
Turan N B, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 104329 (2020)
Development of a new temperature-controlled liquid phase microextraction using deep eutectic solvent for extraction and preconcentration of diazinon, metalaxyl, bromopropylate, oxadiazon, and fenazaquin pesticides from fruit juice and vegetable samples followed by gas chromatography-flame ionization detection
Farajzadeh M A, et al.
Journal of food composition and analysis, 66, 90-97 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service