Skip to Content
Merck
All Photos(1)

Key Documents

843310

TOYOPEARL® LABPAK Bulk Media Sampler

LABPAK HMW Size Exclusion Resin Sampler, pkg of 3 × 150 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
particle size:
45 μm
matrix:
fully porous particle

product line

TOYOPEARL®

form

slurry

packaging

pkg of 3 × 150 mL

technique(s)

HPLC: suitable

matrix

fully porous particle

particle size

45 μm

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

LabPak products are multi-milliliter containers of TOYOPEARL and TSKgel bulk media products. Typically they contain 3 or 4 different ligand types offered for a particular chromatography mode. They are useful for developmental engineers who wish to familiarize themselves with resin physical properties in different buffer systems. The larger resin amounts in LabPak products allow the packing of wider ID and longer columns than available in the ToyoScreen products. This helps the developmental chemist or engineer to better measure under actual packing conditions such properties as dynamic binding capacity, selectivity and column efficiency.

Application

TOYOPEARL® Size Exclusion LABPAK Samplers allow the use of several TOYOPEARL® resins to determine which works best for a particular application. TOYOPEARL® resins are used in separation media and resins.

Components

SEC Media: HW-55F, HW-65F, HW75F (high molecular weight range)

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

102.2 °F

Flash Point(C)

39 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tim Blaschke et al.
Journal of chromatography. A, 1218(29), 4720-4726 (2011-06-22)
Adsorption of native as well as mono-, di-, and tri-PEGylated lysozyme on Toyopearl Gigacap S-650M in sodium phosphate buffer is studied by isothermal titration calorimetry and by independent adsorption equilibrium measurements at pH 6 and 25°C. The production and separation
Marina Urmann et al.
mAbs, 2(4), 395-404 (2010-06-19)
A novel cation-exchange resin, Eshmuno™ S, was compared to Fractogel® SO3(-) (M) and Toyopearl GigaCap S-650M. The stationary phases have different base matrices, and carry specific types of polymeric surface modifications. Three monoclonal antibodies (mAbs) were used as model proteins
Leonard Wt Fweja et al.
The Journal of dairy research, 77(3), 357-367 (2010-06-26)
Lactoperoxidase (LP) was isolated from whey protein by cation-exchange using Carboxymethyl resin (CM-25C) and Sulphopropyl Toyopearl resin (SP-650C). Both batch and column procedures were employed and the adsorption capacities and extraction efficiencies were compared. The resin bed volume to whey

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service