Skip to Content
Merck
All Photos(1)

Key Documents

817354

TSKgel® G2500HHR (5 µm) HPLC Columns

L x I.D. 30 cm × 7.8 mm, polystyrene-divinylbenzene phase

Synonym(s):

TSKgel® HHR Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Pricing and availability is not currently available.

product name

TSKgel® G2500HHR HPLC Column, phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 5 μm particle size

material

stainless steel

Agency

suitable for USP L21

product line

TSKgel®

manufacturer/tradename

Tosoh 17354

parameter

140 °C temperature
2 mL/min flow rate
50 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

matrix

polymer particle platform

matrix active group

polystyrene-divinylbenzene phase

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
817357817356817352
separation technique

size exclusion (SEC)

separation technique

size exclusion (SEC)

separation technique

size exclusion (SEC)

separation technique

size exclusion (SEC)

particle size

5 μm

particle size

5 μm

particle size

5 μm

particle size

5 μm

matrix active group

polystyrene-divinylbenzene phase

matrix active group

polystyrene-divinylbenzene phase

matrix active group

polystyrene-divinylbenzene phase

matrix active group

polystyrene-divinylbenzene phase

pore size

30 Å

pore size

650 Å

pore size

200 Å

pore size

15 Å

agency

suitable for USP L21

agency

suitable for USP L21

agency

suitable for USP L21

agency

suitable for USP L21

General description

TSKgel G2500HHR columns are packed with rigid, porous, polymer beads. They are used for the non-aqueous separation of oligomers with molecular weights less than 20,000 Da. TSKgel G2500HHR columns are shipped in THF, which can be exchanged for a wide variety of organic solvents.
The TSKgel HHR column line consists of eight columns with different pore sizes, TSKgel G1000HHR through TSKgel G7000HHR, and five columns with an extended linear range of the calibration curve. All TSKgel HHR-type columns are shipped in THF, which can be exchanged for a much broader range of organic solvents than TSKgel HXL columns, although Hxl columns are more inert. The maximum operating temperature for most TSKgel HHR-type columns is 140°C, except for GMHHR columns, which can only be used up to 80°C. TSKgel HHR-type columns are conventional GPC columns with dimensions of 7.8mm ID x 30cm.

Compatibility

THF can be replaced with acetone, ethanol, quinoline, benzene, o-dichlorobenzene, ethyl acetate, dodecane, FC-113, carbon tetrachloride, dichloromethane, dichloroethane, trichloroethane, n-hexane, cyclohexane, xylene, tetrahydrofuran, chloroform, 1,4-dioxane, hexafluoroisopropanol, toluene, 1-chloronaphthalene, dimethylaceto acetamide, methyl ethyl ketone, trichlorobenzene, m-cresol, dimethylformamide, methylpyrrolidone, o-chlorophenol/chloroform, dimethyl sulfoxide, pyridine

Physical form

Shipped in THF.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service