Skip to Content
Merck
All Photos(1)

Key Documents

816135

TSKgel® G2500HXL (5 µm) HPLC Columns

L x I.D. 30 cm × 7.8 mm, polystyrene-divinylbenzene phase

Synonym(s):

TSKgel® G1000HXL HPLC Column for Light Scattering Applications

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
L × I.D.:
30 cm × 7.8 mm
particle size:
5 μm
matrix active group:
polystyrene-divinylbenzene phase
pore size:
30 Å
matrix:
polymer particle platform

Product Name

TSKgel® G2500HXL HPLC Column, phase polystyrene-divinylbenzene, L × I.D. 30 cm × 7.8 mm, 5 μm particle size

material

stainless steel

product line

TSKgel®

manufacturer/tradename

Tosoh 16135

parameter

1.2 mL/min flow rate
50 bar max. pressure
60 °C temperature

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

matrix

polymer particle platform

matrix active group

polystyrene-divinylbenzene phase

particle size

5 μm

pore size

30 Å

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

TSKgel G2500Hxl columns are packed with rigid, porous, polymer beads. They are applied to the non-aqueous separation of oligomers and polymers of less than 20,000 Da. As the number of solvents that can be exchanged directly with the shipping solvent (THF) is limited, use TSKgel Hhr-type columns when a broader range of solvents is needed.

Compatibility

THF can be replaced with benzene, chloroform, toluene, xylene, dichloromethane, dichloroethane

Physical form

Shipped in THF.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service