Skip to Content
Merck
All Photos(1)

Key Documents

69135

Supelco

Diisobutyl fumarate

analytical standard

Synonym(s):

Fumaric acid diisobutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2O2CCH=CHCO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
228.28
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.21

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.443 (lit.)

bp

249 °C (lit.)

mp

8 °C (lit.)

density

0.978 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

CC(C)COC(=O)\C=C\C(=O)OCC(C)C

InChI

1S/C12H20O4/c1-9(2)7-15-11(13)5-6-12(14)16-8-10(3)4/h5-6,9-10H,7-8H2,1-4H3/b6-5+

InChI key

RSRICHZMFPHXLE-AATRIKPKSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service