Skip to Content
Merck
All Photos(1)

Key Documents

59553

Supelco

SUPELCOSIL LC-8-DB (5 µm) HPLC Columns

L × I.D. 2 cm × 4 mm Supelguard Guard Cartridge pkg of 2 ea, Guard Cartridge holder required for use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

SUPELCOSIL LC-8-DB Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm

Agency

suitable for USP L7

Quality Level

packaging

pkg of 1 kit

technique(s)

HPLC: suitable

L × I.D.

2 cm × 4 mm

matrix active group

C8 (octyl) phase

particle size

5 μm

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Footnote

Kit includes one cartridge, a stand-alone holder, a piece of tubing, and 2 nuts and ferrules.

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vesna Cerkvenik-Flajs et al.
Analytica chimica acta, 663(2), 165-171 (2010-03-09)
An analytical method has been developed for the simultaneous determination of the following endectocide drugs in milk: ivermectin, abamectin, doramectin, moxidectin, eprinomectin, emamectin and nemadectin. Samples were extracted with acetonitrile, purified with solid-phase extraction on a reversed phase C(8), derivatised
J H Nichols et al.
Clinical chemistry, 40(7 Pt 1), 1312-1316 (1994-07-01)
This automated assay determines the concentration of the antidepressant fluoxetine (Prozac) and its active metabolite norfluoxetine in serum by reversed-phase HPLC with spectrophotometric detection. Extraction, injection, and quantification are performed by the Gilson Aspec automated sample handler. The patient's specimen
Jadwiga Piwowarska et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 805(1), 1-5 (2004-04-29)
A high-performance liquid chromatographic method is described for determination of lidocaine (2-(dietyloamino)-N-(2,6-dimetylofenylo) acetamid) and its metabolite, monoethylglycine xylidide (MEGX), in human serum containing various concentration of bilirubin. Lidocaine and its metabolite were extracted from human serum using dichloromethane. After separation
K Ary et al.
Journal of pharmaceutical and biomedical analysis, 26(2), 179-187 (2001-07-27)
A reversed-phase high-performance liquid chromatographic method with coulometric and UV detection has been developed for the simultaneous determination of morphine, morphine-3-glucuronide and morphine-6-glucuronide. The separation was carried out by using a Supelcosil LC-8 DB reversed-phase column and 0.1 M potassium
P K Kunicki
Journal of chromatography. B, Biomedical sciences and applications, 750(1), 41-49 (2001-02-24)
A HPLC-UV determination of clobazam and N-desmethylclobazam in human serum and urine is presented. After simple liquid-liquid extraction with dichloromethane the compounds and an internal standard diazepam were separated on a Supelcosil LC-8-DB column at ambient temperature under isocratic conditions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service