Skip to Content
Merck
All Photos(1)

Key Documents

58221-U

Supelco

SUPELCOSIL LC-CN (5 µm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, HPLC Column

Synonym(s):

MTO-SUPELCOSIL LC-CN 5UM 15CMX4.6MM HPLC COLUMN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

SUPELCOSIL LC-CN HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm

material

stainless steel column

Agency

suitable for USP L10

product line

SUPELCOSIL

feature

endcapped

packaging

1 ea of

extent of labeling

4% Carbon loading

parameter

≤70 °C temp. range

technique(s)

HPLC: suitable

L × I.D.

15 cm × 4.6 mm

surface area

170 m2/g

surface coverage

surface coverage 3.5 μmol/m2

matrix

silica gel, spherical particle platform
fully porous particle

matrix active group

cyano phase

particle size

5 μm

pore size

120 Å

pH range

2-7.5

separation technique

hydrophilic interaction (HILIC)
normal phase
reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

The LC-CN phase is often used as a substitute for silica because it offers the advantages of a bonded phase (such as quick equilibration, and less sensitivity to small changes of the water content in the mobile phase). More often, however, the LC-CN column is operated under reversed-phase mobile phase conditions.

Application


  • Validated Simple HPLC-UV Method for Mycophenolic Acid (MPA) Monitoring in Human Plasma. Internal Standardization: : This study presents a validated HPLC-UV method for monitoring mycophenolic acid in human plasma. The research highlights the necessity and efficiency of internal standardization in achieving accurate measurements. (Kunicki PK et al., 2021).

  • An Improved HPLC Method for Therapeutic Drug Monitoring of Daunorubicin, Idarubicin, Doxorubicin, Epirubicin, and Their 13-dihydro Metabolites in Human Plasma.: This paper describes an enhanced HPLC method for monitoring therapeutic levels of several chemotherapeutic agents and their metabolites in human plasma, improving the accuracy and reliability of drug monitoring. (Fogli S et al., 1999).

  • Analysis of Diltiazem and Desacetyldiltiazem in Serum by High-Performance Liquid Chromatography.: This research focuses on an HPLC method for analyzing diltiazem and its primary metabolite in serum, providing a robust tool for clinical pharmacokinetic studies. (Paczkowski D et al., 1995).

  • High-Performance Liquid Chromatographic Method for the Determination of Eclanamine and Its N-Desmethyl and N,N-Didesmethyl Metabolites in Urine.: The study details a high-performance liquid chromatographic method for determining eclanamine and its metabolites in urine, crucial for pharmacokinetic and toxicological analysis. (Lakings DB et al., 1988).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

guard cartridge


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Fogli et al.
Therapeutic drug monitoring, 21(3), 367-375 (1999-06-12)
A single high-performance liquid chromatography (HPLC) method, suitable for the analysis of daunorubicin, idarubicin, doxorubicin, epirubicin, and their 13-dihydro metabolites is validated in the present study. Preparation of plasma samples was performed by a first extraction of analytes with a
D Paczkowski et al.
Polish journal of pharmacology, 47(5), 429-434 (1995-09-01)
This paper describes a simple high-performance liquid chromatographic (HPLC) method for the determination of diltiazem and desacetyldiltiazem in human serum. After basic methyl-tert-butyl ether extraction and back-extraction with hydrochloric acid, the drug and its metabolite was injected into a Supelcosil
H Kataoka et al.
Journal of analytical toxicology, 24(4), 257-265 (2000-06-29)
A simple and rapid method for the determination of amphetamine, methamphetamine, and their 3,4-methylenedioxy derivatives in urine samples was developed using automated in-tube solid-phase microextraction (SPME) coupled with liquid chromatography-electrospray ionization mass spectrometry (LC-ESI-MS). In-tube SPME is an extraction technique
H Kataoka et al.
Journal of chromatography. B, Biomedical sciences and applications, 731(2), 353-359 (1999-10-08)
The technique of automated in-tube solid-phase microextraction (SPME) coupled with liquid chromatography-electrospray ionization mass spectrometry (LC-ESI-MS) was evaluated for the determination of ranitidine. In-tube SPME is an extraction technique for organic compounds in aqueous samples, in which analytes are extracted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service